siRNA Details
Gene Symbol: Olfr812 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146795.1 | NP_667006.1 | 1 | 793 | ||
GenBank mRNA | BC132194.1 | - | 1 | 815 | 40130471 | |
BC132196.1 | - | 1 | 815 | 40130473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
