siRNA Details
Gene Symbol: Olfr1156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146817.2 | NP_667028.2 | 1 | 840 | ||
GenBank mRNA | BC116852.1 | - | 1 | 840 | 40126171 | |
BC116854.1 | - | 1 | 840 | 40126173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
