siRNA Details
Gene Symbol: Olfr273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146824.1 | NP_667035.1 | 1 | 778 | ||
GenBank mRNA | BC125463.1 | - | 1 | 839 | 40129978 | |
BC125465.1 | - | 1 | 839 | 40129980 |
