siRNA Details
Gene Symbol: Olfr1220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146900.2 | NP_667111.2 | 1 | 568 | ||
GenBank mRNA | BC125363.1 | - | 1 | 568 | 40129878 | |
BC131900.1 | - | 1 | 568 | 40130177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
