siRNA Details
Gene Symbol: Olfr1282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146907.2 | NP_667118.2 | 1 | 656 |
Gene Symbol: Olfr1283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207236.1 | NP_997119.1 | 1 | 656 | ||
GenBank mRNA | BC125536.1 | - | 1 | 699 | 40130051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
