siRNA Details
Gene Symbol: Olfr1348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146913.1 | NP_667124.1 | 1 | 663 | ||
GenBank mRNA | BC120570.1 | - | 1 | 759 | 40129493 | |
BC137836.1 | - | 1 | 759 | 8860855 |
Gene Symbol: Olfr1347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146385.1 | NP_666497.1 | 1 | 684 | ||
GenBank mRNA | BC128003.1 | - | 1 | 688 | 40135936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
