siRNA Details
Gene Symbol: Olfr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146923.2 | NP_667134.2 | 2 | 740 | ||
GenBank mRNA | AY053400.1 | - | 1 | 244 | ||
BC138172.1 | - | 1 | 643 | 8861191 | ||
BC138173.1 | - | 1 | 643 | 8861192 | ||
X89692.1 | - | 1 | 249 |
Gene Symbol: Olfr376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172686.1 | NP_001166157.1 | 3 | 888 | ||
NM_146922.2 | NP_667133.1 | 2 | 757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
