siRNA Details
Gene Symbol: PSMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102667.1 | NP_001096137.1 | 8 | 655 | ||
NM_001102668.1 | NP_001096138.1 | 7 | 568 | |||
NM_002789.4 | NP_002780.1 | 8 | 731 | |||
GenBank mRNA | AK055714.1 | - | 5 | 2568 | ||
AK315526.1 | - | 8 | 692 | |||
BC005361.1 | - | 8 | 709 | 3685931 | ||
BC022445.1 | - | 8 | 663 | 4287236 | ||
BC022817.2 | - | 6 | 570 | |||
BC047667.1 | - | 8 | 642 | 6060404 | ||
BC093069.1 | - | 7 | 500 | 30414795 | ||
BG616186.1 | - | 7 | 612 | |||
BT009784.1 | - | 6 | 573 | |||
CN337296.1 | - | 7 | 512 | |||
D00763.1 | - | 8 | 632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
