siRNA Details
Gene Symbol: Olfr340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146951.1 | NP_667162.1 | 1 | 393 | ||
GenBank mRNA | BC109141.1 | - | 1 | 393 | 40041785 | |
BC109142.1 | - | 1 | 392 |
Gene Symbol: Olfr345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146945.1 | NP_667156.1 | 1 | 393 | ||
GenBank mRNA | BC148219.1 | - | 1 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
