siRNA Details
Gene Symbol: PSMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102667.1 | NP_001096137.1 | 4 | 165 | ||
NM_002789.4 | NP_002780.1 | 4 | 241 | |||
GenBank mRNA | AK055714.1 | - | 1 | 2078 | ||
AK315526.1 | - | 4 | 202 | |||
AL110100.1 | - | 4 | 179 | |||
BC005361.1 | - | 4 | 219 | 3685931 | ||
BC022445.1 | - | 4 | 173 | 4287236 | ||
BC022817.2 | - | 2 | 80 | |||
BC047667.1 | - | 4 | 152 | 6060404 | ||
BC056249.1 | - | 4 | 245 | 4093514 | ||
BG616186.1 | - | 3 | 121 | |||
BT009784.1 | - | 2 | 83 | |||
BU930105.1 | - | 133 | ||||
D00763.1 | - | 4 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
