siRNA Details
Gene Symbol: Olfr1242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146968.2 | NP_667179.1 | 1 | 673 | ||
GenBank mRNA | BC120865.1 | - | 1 | 673 | 40129788 | |
BC125386.1 | - | 1 | 673 | 40129901 |
Gene Symbol: Olfr1243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146969.1 | NP_667180.1 | 1 | 637 | ||
GenBank mRNA | BC111812.1 | - | 1 | 632 |

More Information
Other Species Targeted

Gene Ontology Categories:
