siRNA Details
Gene Symbol: PSMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002792.3 | NP_002783.1 | 6, 7 | 802 | ||
GenBank mRNA | AF022815.1 | - | 6, 7 | 671 | ||
AF054185.1 | - | 6, 7 | 732 | |||
AK312025.1 | - | 6, 7 | 697 | |||
BC001895.1 | - | 6, 7 | 735 | |||
BC004427.1 | - | 6, 7 | 720 | 2819923 | ||
BC014999.1 | - | 6, 7 | 730 | |||
BG575009.1 | - | 6, 7 | 796 | |||
BT007165.1 | - | 6, 7 | 647 | |||
GQ472224.1 | - | 6, 7 | 720 |
