siRNA Details
Gene Symbol: Olfr399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147004.2 | NP_667215.2 | 1 | 594 | ||
GenBank mRNA | BC120820.1 | - | 1 | 594 | 40129743 | |
BC120846.1 | - | 1 | 594 | 40129769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
