siRNA Details
Gene Symbol: Olfr1353 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147042.2 | NP_667253.1 | 3 | 434 | ||
GenBank mRNA | BC119112.1 | - | 1 | 183 | 8733861 | |
BC120824.1 | - | 1 | 183 | 40129747 |
