siRNA Details
Gene Symbol: Olfr668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147059.1 | NP_667270.1 | 1 | 916 |
Gene Symbol: Olfr666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147096.1 | NP_667307.1 | 1 | 916 |

More Information
Other Species Targeted

Gene Ontology Categories:
