siRNA Details
Gene Symbol: Olfr686 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147069.1 | NP_667280.1 | 1 | 114 | ||
GenBank mRNA | BC132004.1 | - | 1 | 149 | 40130281 | |
BC132006.1 | - | 1 | 149 | 40130283 |

More Information
Other Species Targeted

Gene Ontology Categories:
