siRNA Details
Gene Symbol: Olfr619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147076.2 | NP_667287.2 | 1 | 915 | ||
GenBank mRNA | BC141053.1 | - | 1 | 915 | 9055665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
