siRNA Details
Gene Symbol: Olfr561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147092.1 | NP_667303.1 | 1 | 422 | ||
GenBank mRNA | BC120569.1 | - | 1 | 453 | 40129492 | |
BC120571.1 | - | 1 | 453 | 40129494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
