siRNA Details
Gene Symbol: Olfr628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147097.2 | NP_667308.2 | 1 | 216 | ||
GenBank mRNA | BC109148.1 | - | 1 | 302 | 40046009 | |
BC109149.1 | - | 1 | 302 | 40046010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
