siRNA Details
Gene Symbol: Olfr630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147098.2 | NP_667309.1 | 2 | 981 | ||
GenBank mRNA | BC116998.1 | - | 1 | 939 | 40126317 | |
BC117000.1 | - | 1 | 939 | 40126319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
