siRNA Details
Gene Symbol: Olfr644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147121.1 | NP_667332.1 | 1 | 398 | ||
GenBank mRNA | BC099886.1 | - | 1 | 390 | ||
BC099887.1 | - | 1 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
