siRNA Details
Gene Symbol: Klk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008940.2 | NP_032966.1 | 4 | 1001 | ||
GenBank mRNA | AB074296.1 | - | 2 | 263 | ||
BC055895.1 | - | 4 | 515 | 4019947 | ||
D30785.1 | - | 4 | 1001 |

More Information
Other Species Targeted

Gene Ontology Categories:
