siRNA Details
Gene Symbol: Trim61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177551.1 | NP_001171022.1 | 2 | 1392 | ||
NM_153110.4 | NP_694750.3 | 3 | 1465 | |||
GenBank mRNA | AK054544.1 | - | 2 | 1278 | ||
AY063495.1 | - | 2 | 1227 | |||
BC064738.1 | - | 3 | 1343 | 30049322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
