siRNA Details
Gene Symbol: Cadm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153112.3 | NP_694752.1 | 4 | 564 | ||
GenBank mRNA | AK143204.1 | - | 4 | 565 | ||
AY059394.1 | - | 4 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
