siRNA Details
Gene Symbol: Otos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153114.2 | NP_694754.1 | 5 | 676 | ||
GenBank mRNA | AK083090.1 | - | 4 | 2964 | ||
AY078071.1 | - | 4 | 424 | |||
BC100323.1 | - | 5 | 676 | 30788363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
