siRNA Details
Gene Symbol: Prss45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153172.1 | NP_694812.1 | 1, 2 | 160 | ||
GenBank mRNA | AF392062.1 | - | 1, 2 | 160 | ||
AK077121.1 | - | 1, 2 | 158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
