siRNA Details
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029926.5 | NP_084202.2 | 4 | 577 | ||
GenBank mRNA | AF445803.1 | - | 4 | 565 | ||
AK020397.2 | - | 4 | 586 | |||
AK028837.1 | - | 4 | 518 | |||
AK029028.1 | - | 4 | 578 | |||
AK139992.1 | - | 566 | ||||
BC051676.1 | - | 4 | 570 | 30017484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
