siRNA Details
Gene Symbol: Rps15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170669.2 | NP_733769.1 | 1 | 121 | ||
GenBank mRNA | AK008350.1 | - | 2 | 132 | ||
AK010652.1 | - | 2 | 162 | |||
AK011514.1 | - | 2 | 135 | |||
AK012485.1 | - | 2 | 139 | |||
AK012618.1 | - | 2 | 135 | |||
AK019235.1 | - | 2 | 135 | |||
AK028135.1 | - | 2 | 135 | |||
AK032522.1 | - | 2 | 161 | |||
BC051205.1 | - | 1 | 130 | |||
BC054792.1 | - | 1 | 121 | 6832855 | ||
BC055697.1 | - | 1 | 116 | 6411145 | ||
BC076563.1 | - | 2 | 126 | 6411321 | ||
BC081428.1 | - | 1 | 116 | 6412149 | ||
BC086885.1 | - | 2 | 134 | 6763031 | ||
BC087867.1 | - | 2 | 137 | 6543067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
