siRNA Details
Gene Symbol: Kcnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025581.1 | NP_001020752.1 | 3 | 903 | ||
GenBank mRNA | AK042937.1 | - | 3 | 323 | ||
AK045425.1 | - | 3 | 905 | |||
BC116289.1 | - | 3 | 905 | 40094886 | ||
BC116290.1 | - | 3 | 904 | 40094891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
