siRNA Details
Gene Symbol: Ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172300.3 | NP_758504.3 | 5 | 828 | ||
GenBank mRNA | AK005524.1 | - | 3 | 379 | ||
AK077793.1 | - | 5 | 827 | |||
AK149770.1 | - | 5 | 828 | |||
AK152529.1 | - | 5 | 542 | |||
AK153328.1 | - | 5 | 559 | |||
AK168942.1 | - | 5 | 521 | |||
BC027266.1 | - | 5 | 453 | |||
BC054412.1 | - | 5 | 531 | 2646768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
