siRNA Details
Gene Symbol: Greb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252071.1 | NP_001239000.1 | 20 | 3447 | ||
NM_015764.4 | NP_056579.2 | 22 | 3853 | |||
GenBank mRNA | AB093248.1 | - | 5 | 830 | ||
AF180470.1 | - | 21 | 3509 | |||
AK147337.1 | - | 20 | 3381 | |||
AK157808.1 | - | 20 | 3530 | |||
BC160931.1 | - | 21 | 3379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
