siRNA Details
Gene Symbol: Greb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252071.1 | NP_001239000.1 | 11 | 1956 | ||
NM_015764.4 | NP_056579.2 | 13 | 2278 | |||
GenBank mRNA | AF180470.1 | - | 12 | 1934 | ||
AK147337.1 | - | 11 | 1806 | |||
AK157808.1 | - | 11 | 1890 | |||
BC160931.1 | - | 12 | 1888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
