siRNA Details
Gene Symbol: Egflam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289496.1 | NP_001276425.1 | 6 | 928 | ||
NM_001289498.1 | NP_001276427.1 | 6 | 923 | |||
NM_178748.6 | NP_848863.1 | 6 | 928 | |||
GenBank mRNA | AK031136.1 | - | 6 | 837 | ||
AK033332.1 | - | 6 | 845 | |||
AK037223.1 | - | 6 | 830 | |||
AK041546.1 | - | 6 | 809 | |||
AK087659.1 | - | 6 | 808 | |||
AK158909.1 | - | 6 | 924 | |||
BC095994.1 | - | 6 | 658 | 6467602 | ||
BC150710.1 | - | 6 | 648 | 9087621 | ||
DQ223720.1 | - | 6 | 632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
