siRNA Details
Gene Symbol: Mroh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033365.2 | NP_001028537.1 | 6 | 621 | ||
GenBank mRNA | AK132992.1 | - | 1048 | |||
AK158799.1 | - | 6 | 622 | |||
BC150895.1 | - | 6 | 623 | 9087806 | ||
BC172046.1 | - | 6 | 623 | 9054332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
