siRNA Details
Gene Symbol: Abat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170978.1 | NP_001164449.1 | 3, 4 | 368 | ||
NM_172961.3 | NP_766549.2 | 3, 4 | 368 | |||
GenBank mRNA | AK036128.1 | - | 3, 4 | 313 | ||
AK132282.1 | - | 3, 4 | 317 | |||
AK160802.1 | - | 3, 4 | 316 | |||
AK165348.1 | - | 3, 4 | 318 | |||
BC058079.1 | - | 3, 4 | 308 | 6826741 | ||
BC058521.1 | - | 3, 4 | 308 | 6825487 | ||
CX237932.1 | - | 3, 4 | 368 |
