siRNA Details
Gene Symbol: Ss18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161369.1 | NP_001154841.1 | 7 | 1041 | ||
NM_001161370.1 | NP_001154842.1 | 6 | 819 | |||
NM_001161371.1 | NP_001154843.1 | 6 | 819 | |||
NM_009280.2 | NP_033306.2 | 7 | 1041 | |||
GenBank mRNA | AK039430.1 | - | 7 | 891 | ||
AK051601.1 | - | 672 | ||||
AK052476.1 | - | 6 | 673 | |||
AK083622.1 | - | 7 | 903 | |||
AK149325.1 | - | 7 | 903 | |||
AK166053.1 | - | 7 | 894 | |||
AK179801.1 | - | 1 | 96 | |||
BC065062.1 | - | 7 | 878 | |||
BC096742.1 | - | 7 | 878 | 6416415 | ||
X93357.1 | - | 7 | 1041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
