siRNA Details
Gene Symbol: PSMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002817.3 | NP_002808.3 | 4, 5 | 504 | ||
NM_175932.2 | NP_787128.2 | 2, 3 | 510 | |||
XM_011520235.2 | XP_011518537.1 | 4, 5 | 637 | |||
GenBank mRNA | AB009398.1 | - | 4, 5 | 331 | ||
AF083245.1 | - | 4, 5 | 316 | |||
AF086708.1 | - | 4, 5 | 262 | |||
AF107837.1 | - | 4, 5 | 295 | |||
AK094775.1 | - | 2, 3 | 341 | |||
AK222895.1 | - | 4, 5 | 335 | |||
AK295944.1 | - | 3, 4 | 301 | |||
AK307447.1 | - | 3, 4 | 229 | |||
AK314733.1 | - | 4, 5 | 766 | |||
BC001100.2 | - | 4, 5 | 317 | 3506530 | ||
BC001747.1 | - | 4, 5 | 285 | 3535807 | ||
BM456586.1 | - | 4, 5 | 335 | |||
BT007307.1 | - | 4, 5 | 262 | |||
BX647314.1 | - | 3, 4 | 239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
