siRNA Details
Gene Symbol: Zfp608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175751.4 | NP_786927.2 | 5 | 3037 | ||
GenBank mRNA | AK077030.1 | - | 4 | 1764 | ||
BC059268.1 | - | 4 | 2704 | |||
BC092219.1 | - | 5 | 2770 | 6830664 | ||
BC100388.1 | - | 1 | 286 | |||
BC151073.1 | - | 4 | 2556 | 9087986 | ||
BC151075.1 | - | 4 | 2556 | 9087988 |
