siRNA Details
Gene Symbol: 4930503L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172967.2 | NP_766555.1 | 3 | 811 | ||
GenBank mRNA | AK037066.1 | - | 3 | 812 | ||
AK080780.1 | - | 3 | 842 | |||
AK087788.1 | - | 3 | 784 | |||
AK148410.1 | - | 3 | 835 | |||
AK160248.1 | - | 3 | 685 | |||
AK160714.1 | - | 3 | 781 | |||
BC049726.1 | - | 323 | ||||
BC057927.1 | - | 2 | 528 | 5290837 | ||
HQ438973.1 | - | 2 | 332 | |||
HQ438974.1 | - | 2 | 332 | |||
HQ438975.1 | - | 2 | 332 | |||
HQ438978.1 | - | 1 | 291 | |||
HQ438984.1 | - | 1 | 291 | |||
HQ438986.1 | - | 1 | 291 | |||
HQ438990.1 | - | 1 | 291 | |||
HQ438991.1 | - | 1 | 291 | |||
HQ438993.1 | - | 1 | 291 | |||
HQ438995.1 | - | 1 | 291 | |||
HQ439007.1 | - | 1 | 291 | |||
HQ439008.1 | - | 1 | 291 | |||
HQ439010.1 | - | 1 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
