siRNA Details
Gene Symbol: PSMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002817.3 | NP_002808.3 | 7 | 754 | ||
NM_175932.2 | NP_787128.2 | 5 | 760 | |||
XM_011520235.2 | XP_011518537.1 | 7 | 887 | |||
GenBank mRNA | AB009398.1 | - | 7 | 581 | ||
AF083245.1 | - | 7 | 566 | |||
AF086708.1 | - | 7 | 512 | |||
AF107837.1 | - | 7 | 545 | |||
AK094775.1 | - | 5 | 591 | |||
AK222895.1 | - | 7 | 585 | |||
AK295944.1 | - | 6 | 551 | |||
AK307447.1 | - | 6 | 479 | |||
AK314733.1 | - | 7 | 1016 | |||
BC001100.2 | - | 7 | 567 | 3506530 | ||
BC001747.1 | - | 7 | 535 | 3535807 | ||
BM456586.1 | - | 7 | 585 | |||
BT007307.1 | - | 7 | 512 | |||
BX647314.1 | - | 6 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
