siRNA Details
Gene Symbol: PSMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF083245.1 | - | 4 | 293 | ||
AF086708.1 | - | 4 | 239 | |||
AF107837.1 | - | 4 | 272 | |||
AK295944.1 | - | 3 | 278 | |||
AK307447.1 | - | 3 | 206 | |||
BC001100.2 | - | 4 | 294 | 3506530 | ||
BC001747.1 | - | 4 | 262 | 3535807 | ||
BM456586.1 | - | 4 | 312 | |||
BT007307.1 | - | 4 | 239 | |||
BX647314.1 | - | 3 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
