siRNA Details
Gene Symbol: Atp5h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027862.1 | NP_082138.1 | 3 | 239 | ||
GenBank mRNA | AF354051.1 | - | 2 | 232 | ||
AK002376.1 | - | 3 | 239 | |||
AK088617.1 | - | 2 | 234 | |||
BC016547.1 | - | 3 | 237 | 2631309 | ||
BC081431.1 | - | 3 | 238 | 6408035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
