siRNA Details
Gene Symbol: Atp5h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027862.1 | NP_082138.1 | 3 | 171 | ||
GenBank mRNA | AF354051.1 | - | 2 | 164 | ||
AK002376.1 | - | 3 | 171 | |||
AK088617.1 | - | 2 | 166 | |||
BC016547.1 | - | 3 | 169 | 2631309 | ||
BC081431.1 | - | 3 | 170 | 6408035 |

More Information
Other Species Targeted

Gene Ontology Categories:
