siRNA Details
Gene Symbol: Dpp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199021.3 | NP_950186.3 | 17, 18 | 1653 | ||
GenBank mRNA | AK046842.1 | - | 17, 18 | 1654 | ||
BC029696.1 | - | 2, 3 | 135 | |||
BC063074.1 | - | 7, 8 | 575 | 5697531 | ||
BC067026.1 | - | 17, 18 | 1646 | 6853947 |
