siRNA Details
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183392.2 | NP_899248.1 | 4, 5 | 661 | ||
GenBank mRNA | AK087827.1 | - | 4, 5 | 555 | ||
AK088855.1 | - | 4, 5 | 586 | |||
AK133841.1 | - | 4, 5 | 659 | |||
AK150182.1 | - | 4, 5 | 586 | |||
AK150841.1 | - | 4, 5 | 547 | |||
AK151896.1 | - | 4, 5 | 547 | |||
AK152136.1 | - | 4, 5 | 547 | |||
AK162037.1 | - | 4, 5 | 593 | |||
BC044303.1 | - | 4, 5 | 589 | |||
BC068114.1 | - | 4, 5 | 546 | 5707888 |
