siRNA Details
Gene Symbol: Fam171a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081161.1 | NP_001074630.1 | 3 | 547 | ||
GenBank mRNA | AK032237.1 | - | 3 | 523 | ||
AK036270.1 | - | 3 | 325 | |||
AK077831.1 | - | 3 | 534 | |||
BC138175.1 | - | 3 | 466 | 8861194 | ||
BC145171.1 | - | 3 | 466 |
