siRNA Details
Gene Symbol: Ccdc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199310.2 | NP_955014.2 | 3 | 469 | ||
GenBank mRNA | AK040670.1 | - | 3 | 439 | ||
AK132604.1 | - | 3 | 461 | |||
AK133813.1 | - | 3 | 443 | |||
AK147152.1 | - | 3 | 445 | |||
BC023926.1 | - | 3 | 463 | 5324154 | ||
BC052687.1 | - | 3 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
