siRNA Details
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147153.3 | NP_671495.1 | 4 | 336 | ||
NM_178851.3 | NP_849182.1 | 3 | 303 | |||
GenBank mRNA | AB028844.1 | - | 3 | 283 | ||
AF281050.1 | - | 4 | 313 | |||
AF281051.1 | - | 3 | 280 | |||
AK038824.1 | - | 3 | 292 | |||
AK042253.1 | - | 3 | 292 | |||
AK084743.1 | - | 3 | 303 | |||
AK129210.1 | - | 3 | 298 | |||
AK140041.1 | - | 4 | 341 | |||
AK141424.1 | - | 3 | 286 | |||
AK154103.1 | - | 3 | 292 | |||
AK159889.1 | - | 3 | 296 | |||
AK160109.1 | - | 3 | 296 | |||
AK170831.1 | - | 4 | 363 | |||
AK171008.1 | - | 3 | 292 | |||
BB655861.1 | - | 4 | 329 | |||
BC007479.1 | - | 3 | 254 | 3492875 | ||
BY125125.1 | - | 3 | 304 |

More Information
Other Species Targeted

Gene Ontology Categories:
