siRNA Details
Gene Symbol: Fbxo10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024142.1 | NP_001019313.1 | 8 | 2288 | ||
GenBank mRNA | AK142389.1 | - | 7 | 2212 | ||
BC052485.1 | - | 7 | 1755 | |||
BC054731.1 | - | 8 | 2278 | |||
BC062126.1 | - | 7 | 1646 | |||
BC079571.1 | - | 2 | 404 | |||
BC094565.1 | - | 7 | 2072 | 6836944 |
