siRNA Details
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128606.1 | NP_001122078.1 | 2 | 665 | ||
NM_001128607.1 | NP_001122079.1 | 2 | 575 | |||
NM_183428.3 | NP_906273.3 | 2 | 1927 | |||
GenBank mRNA | AB370261.1 | - | 1 | 450 | ||
AK220462.1 | - | 2 | 572 | |||
BC068138.1 | - | 2 | 535 | |||
BC079875.1 | - | 2 | 494 | 6844082 | ||
CA324069.1 | - | 2 | 674 | |||
CB723734.1 | - | 2 | 494 | |||
L00919.1 | - | 2 | 1921 |

More Information
Other Species Targeted

Gene Ontology Categories:
